Perfluorohept-2-ene
Lieferant: Apollo Scientific
PC7688-5G
APOSPC7688-5GEA
172
EUR
APOSPC7688-5G
Perfluorohept-2-ene
Perfluorohept-2-ene
Formel:
C₇F₁₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094437 CAS-Nummer: 1582-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...