N-Formyl-2-brom-4-(trifluormethoxy)anilin 98%
Lieferant: Apollo Scientific
PC7704-1G
APOSPC7704-1GEA
132
EUR
APOSPC7704-1G
N-Formyl-2-brom-4-(trifluormethoxy)anilin 98%
N-Formyl-2-brom-4-(trifluormethoxy)anilin
Formel:
C₈H₅BrF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09475870 CAS-Nummer: 941294-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...