4-(Trifluormethyl)piperidin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

PC7713-1G PC7713-5G PC7713-10G
APOSPC7713-1GEA 21.9 EUR
APOSPC7713-1G APOSPC7713-5G APOSPC7713-10G
4-(Trifluormethyl)piperidin Hydrochlorid
4-(Trifluormethyl)piperidin Hydrochlorid
Formel: C₆H₁₁ClF₃N
Molecular Weight: 189.61 g/mol
Schmelzpunkt: 155…160 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD04971993
CAS-Nummer: 155849-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR