3-(Trifluoromethyl)-L-phenylalanine 97%
Lieferant: Apollo Scientific
PC7714-1G
PC7714-5G
APOSPC7714-1GEA
246
EUR
APOSPC7714-1G
APOSPC7714-5G
3-(Trifluoromethyl)-L-phenylalanine 97%
3-(Trifluoromethyl)-L-phenylalanine
Formel:
C₁₀H₁₀F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044945 CAS-Nummer: 14464-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...