1-(3-(Trifluormethyl)phenyl)imidazol 98%

Lieferant: Apollo Scientific
Achtung

PC7718F-250MG
APOSPC7718F-250MGEA 16.1 EUR
APOSPC7718F-250MG
1-(3-(Trifluormethyl)phenyl)imidazol 98%
1-(3-(Trifluoromethyl)phenyl)imidazole
Formel: C₁₀H₇F₃N₂
Molecular Weight: 212.17 g/mol
Siedepunkt: 70 °C/0.1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00060487
CAS-Nummer: 25371-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR