4-(Trifluoromethyl)-2-pyridinecarbonitrile

Lieferant: Apollo Scientific
Achtung

PC7722-250MG PC7722-1G
APOSPC7722-250MGEA 31.9 EUR
APOSPC7722-250MG APOSPC7722-1G
4-(Trifluoromethyl)-2-pyridinecarbonitrile
4-(Trifluoromethyl)-2-pyridinecarbonitrile
Formel: C₇H₃F₃N₂
Siedepunkt: 58…60 °C/1,5mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD09038236
CAS-Nummer: 936841-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR