4-(Trifluoromethyl)-2-pyridinecarbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethyl)picolinonitrile
PC7722-250MG
PC7722-1G
APOSPC7722-250MGEA
31.9
EUR
APOSPC7722-250MG
APOSPC7722-1G
4-(Trifluoromethyl)-2-pyridinecarbonitrile
4-(Trifluoromethyl)-2-pyridinecarbonitrile
Formel:
C₇H₃F₃N₂ Molecular Weight: 172.11 g/mol Siedepunkt: 58…60 °C/1.5mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD09038236 CAS-Nummer: 936841-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...