4-Amino-7-(trifluormethyl)chinolin

Lieferant: Apollo Scientific
Achtung

PC7756-1G PC7756-250MG PC7756-500MG
APOSPC7756-1GEA 203 EUR
APOSPC7756-1G APOSPC7756-250MG APOSPC7756-500MG
4-Amino-7-(trifluormethyl)chinolin
4-Amino-7-(trifluoromethyl)quinoline
Formel: C₁₀H₇F₃N₂
Molecular Weight: 212.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269411
CAS-Nummer: 243666-11-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR