1-Bromo-2-chloro-3,4,5-trifluorobenzene

Lieferant: Apollo Scientific

PC7759-1G
APOSPC7759-1GEA 425 EUR
APOSPC7759-1G
1-Bromo-2-chloro-3,4,5-trifluorobenzene
1-Bromo-2-chloro-3,4,5-trifluorobenzene
Formel: C₆HBrClF₃
Molecular Weight: 245,424 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070736
CAS-Nummer: 122375-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR