2-(Trifluoromethyl)propene 97%

Lieferant: Apollo Scientific
Gefahr

PC7760-25G
APOSPC7760-25GEA 725 EUR
APOSPC7760-25G
2-(Trifluoromethyl)propene 97%
2-(Trifluoromethyl)propene
Formel: C₄H₅F₃
Molecular Weight: 110,08 g/mol
Siedepunkt: 44383 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042093
CAS-Nummer: 374-00-5
UN: 3161
ADR: 2.1,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR