Trifluor(trifluormethoxy)ethylen 99%

Lieferant: Apollo Scientific
Gefahr

PC7778E-100G PC7778E-25G
APOSPC7778E-100GEA 928 EUR
APOSPC7778E-100G APOSPC7778E-25G
Trifluor(trifluormethoxy)ethylen 99%
Trifluor(trifluormethoxy)ethylen

Improves low temperature performance & chemical resistance when used as a co-polymer to VDF &/or PTFE.

Formel: C₃F₆O
Siedepunkt: –23 °C
Dichte: 1,43 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00221780
CAS-Nummer: 1187-93-5
EINECS: 214-703-7
UN: 3153
ADR: 2.1,

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR