(Trifluormethyl)trimethylsilan 99%

Lieferant: Apollo Scientific
Gefahr

PC7779-5G
APOSPC7779-5GEA 0 EUR
APOSPC7779-5G
(Trifluormethyl)trimethylsilan 99%
(Trifluoromethyl)trimethylsilane

Ruppert's reagent - valuable trifluoromethylating agent of electrophilic compounds.

Formel: C₄H₉F₃Si
Siedepunkt: 54…55 °C
Dichte: 0,962 g/cm³ (25 °C)
Flammpunkt: –17 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00145454
CAS-Nummer: 81290-20-2
UN: 1993
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR