3,6-Difluorphthalsäureanhydrid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,6-Difluor-1,2-benzoldicarbonsäureanhydrid
PC7799-1G
APOSPC7799-1GEA
377
EUR
APOSPC7799-1G
3,6-Difluorphthalsäureanhydrid 97%
3,6-Difluorphthalsäureanhydrid
Formel:
C₈H₂F₂O₃ Schmelzpunkt: 218…221 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134537 CAS-Nummer: 652-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...