3,4-Dihydro-7-fluoro-4-methyl-1H-1,4-benzodiazepine-2,5-dione 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,5-Dioxo-7-fluoro-4-methyl-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine, 3,4-Dihydro-7-fluoro-4-methyl-1H-benzo[e]-1,4-diazepine-2,5-dione, 7-Fluoro-4-methyl-3,4-dihydro-1H-benzo[e][1,4]diazepine-2,5-dione
PC7801-1G
APOSPC7801-1GEA
173
EUR
APOSPC7801-1G
3,4-Dihydro-7-fluoro-4-methyl-1H-1,4-benzodiazepine-2,5-dione 97%
3,4-Dihydro-7-fluoro-4-methyl-1H-1,4-benzodiazepine-2,5-dione
Formel:
C₁₀H₉FN₂O₂ Molecular Weight: 208,189 g/mol Schmelzpunkt: 213…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01063090 CAS-Nummer: 78755-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...