3,3,3-Trifluor-1-propanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,3,3-Trifluoropropan-1-ol
PC7819R-25G
APOSPC7819R-25GEA
135
EUR
APOSPC7819R-25G
3,3,3-Trifluor-1-propanol 97%
3,3,3-Trifluor-1-propanol
Formel:
C₃H₅F₃O Molecular Weight: 114.07 g/mol Siedepunkt: 100…101 °C Dichte: 1.294 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190649 CAS-Nummer: 2240-88-2 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...