Methyl 2-nitro-4-(trifluoromethyl)benzoate 98%

Lieferant: Apollo Scientific
Achtung

PC7828-5G PC7828-1G PC7828-25G
APOSPC7828-5GEA 23.5 EUR
APOSPC7828-5G APOSPC7828-1G APOSPC7828-25G
Methyl 2-nitro-4-(trifluoromethyl)benzoate 98%
Methyl 2-nitro-4-(trifluoromethyl)benzoate
Formel: C₉H₆F₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08282772
CAS-Nummer: 228418-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR