Perfluoro(4-methyl-3,6-dioxaoct-7-ene)sulphonyl fluoride

Lieferant: Apollo Scientific
Achtung

PC7831-1G PC7831-25G PC7831-5G
APOSPC7831-1GEA 55.1 EUR
APOSPC7831-1G APOSPC7831-25G APOSPC7831-5G
Perfluoro(4-methyl-3,6-dioxaoct-7-ene)sulphonyl fluoride
Perfluoro(4-methyl-3,6-dioxaoct-7-ene)sulphonyl fluoride

Intermediate for incorporation of sulphonic acid groups into polymers.

Formel: C₇F₁₄O₄S
Siedepunkt: 135 °C
Dichte: 1,7 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00798138
CAS-Nummer: 16090-14-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR