2,4,6-Trifluorpyrimidin 97%
Lieferant: Apollo Scientific
PC7834-5G
APOSPC7834-5GEA
190
EUR
APOSPC7834-5G
2,4,6-Trifluorpyrimidin 97%
2,4,6-Trifluoropyrimidine
Formel:
C₄HF₃N₂ Molecular Weight: 134,06 g/mol Siedepunkt: 100…102 °C Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039705 CAS-Nummer: 696-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...