3,4-Difluorsalicylsäure 98%

Lieferant: Apollo Scientific
Achtung

PC7835-1G
APOSPC7835-1GEA 87.3 EUR
APOSPC7835-1G
3,4-Difluorsalicylsäure 98%
3,4-Difluorsalicylsäure
Formel: C₇H₄F₂O₃
Molecular Weight: 174.1 g/mol
Schmelzpunkt: 177…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03428564
CAS-Nummer: 189283-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR