3,4-Difluorophenylglycine

Lieferant: Apollo Scientific

PC7856-1G PC7856-5G PC7856-10G
APOSPC7856-1GEA 159 EUR
APOSPC7856-1G APOSPC7856-5G APOSPC7856-10G
3,4-Difluorophenylglycine
3,4-Difluorophenylglycine
Formel: C₈H₇F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06246973
CAS-Nummer: 225641-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR