4,5-Difluoro-2-methylbenzoic acid 98%

Lieferant: Apollo Scientific
Achtung

PC7873-1G PC7873-250MG PC7873-5G
APOSPC7873-1GEA 72.5 EUR
APOSPC7873-1G APOSPC7873-250MG APOSPC7873-5G
4,5-Difluoro-2-methylbenzoic acid 98%
4,5-Difluoro-2-methylbenzoic acid
Formel: C₈H₆F₂O₂
Molecular Weight: 172.13 g/mol
Schmelzpunkt: 117…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08062377
CAS-Nummer: 183237-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR