3-Brom-5-fluor-4-methoxyanilin 98%

Lieferant: Apollo Scientific

PC7874-1G
APOSPC7874-1GEA 143 EUR
APOSPC7874-1G
3-Brom-5-fluor-4-methoxyanilin 98%
3-Brom-5-fluor-4-methoxyanilin
Formel: C₇H₇BrFNO
Molecular Weight: 220,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08062384
CAS-Nummer: 875664-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR