6-Bromo-3-fluoro-o-xylene 97%

Lieferant: Apollo Scientific
Achtung

PC7883-1G
APOSPC7883-1GEA 0 EUR
APOSPC7883-1G
6-Bromo-3-fluoro-o-xylene 97%
6-Bromo-3-fluoro-o-xylene
Formel: C₈H₈BrF
Lagertemperatur: Raumtemperatur
MDL: MFCD03789115
CAS-Nummer: 52548-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR