Methyl-2,2,3,3-tetrafluoropropyl carbonate 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC7886-1G
APOSPC7886-1GEA
69.6
EUR
APOSPC7886-1G
Methyl-2,2,3,3-tetrafluoropropyl carbonate 99%
Methyl-2,2,3,3-tetrafluoropropyl carbonate
Formel:
C₅H₆F₄O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08688580 CAS-Nummer: 156783-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...