Tris(3-fluorphenyl)phosphin 97%

Lieferant: Apollo Scientific
Achtung

PC7886B-1G
APOSPC7886B-1GEA 21.9 EUR
APOSPC7886B-1G
Tris(3-fluorphenyl)phosphin 97%
Tris(3-fluorphenyl)phosphin
Formel: C₁₈H₁₂F₃P
Molecular Weight: 316,26 g/mol
Schmelzpunkt: 62…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015722
CAS-Nummer: 23039-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR