Tris(4-fluorphenyl)phosphin 97%

Lieferant: Apollo Scientific

PC7886C-100G
APOSPC7886C-100GEA 126 EUR
APOSPC7886C-100G
Tris(4-fluorphenyl)phosphin 97%
Tris(4-fluorphenyl)phosphin
Formel: C₁₈H₁₂F₃P
Molecular Weight: 316.26 g/mol
Schmelzpunkt: 80…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013553
CAS-Nummer: 18437-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR