2-Brom-5-(trifluormethyl)benzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC7887-1G PC7887-5G PC7887-10G
APOSPC7887-1GEA 33.5 EUR
APOSPC7887-1G APOSPC7887-5G APOSPC7887-10G
2-Brom-5-(trifluormethyl)benzylbromid 97%
2-Brom-5-(trifluormethyl)benzylbromid
Formel: C₈H₅Br₂F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04973762
CAS-Nummer: 886496-63-5
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR