4-Fluorphthalsäure

Lieferant: Apollo Scientific
Achtung

PC7897-1G
APOSPC7897-1GEA 21.9 EUR
APOSPC7897-1G
4-Fluorphthalsäure
4-Fluorphthalsäure
Formel: C₈H₅FO₄
Molecular Weight: 184.12 g/mol
Schmelzpunkt: 146…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06208213
CAS-Nummer: 320-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR