4-Brom-2,5-difluorphenol 98+%
Lieferant: Apollo Scientific
|
Achtung
|
PC7898-5G
APOSPC7898-5GEA
27.7
EUR
APOSPC7898-5G
4-Brom-2,5-difluorphenol 98+%
4-Brom-2,5-difluorphenol
Formel:
C₆H₃BrF₂O Schmelzpunkt: 44…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973751 CAS-Nummer: 486424-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...