1,1-Dimethyl-2-fluoroethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC7908-1G
APOSPC7908-1GEA 864 EUR
APOSPC7908-1G
1,1-Dimethyl-2-fluoroethylamine hydrochloride
1,1-Dimethyl-2-fluoroethylamine hydrochloride
Formel: C₄H₁₀FN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD07779327
CAS-Nummer: 112433-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR