6,7-Dichlor-3-(trifluormethyl)chinoxalin-2-on 98%

Lieferant: Apollo Scientific

Synonyme: 6,7-Dichloro-3-(trifluoromethyl)quinoxalin-2(1H)-one, 6,7-Dichloro-2-hydroxy-3-(trifluoromethyl)quinoxaline

PC7913-1G
APOSPC7913-1GEA 176 EUR
APOSPC7913-1G
6,7-Dichlor-3-(trifluormethyl)chinoxalin-2-on 98%
6,7-Dichlor-3-(trifluormethyl)chinoxalin-2-on
Formel: C₉H₃Cl₂F₃N₂O
Molecular Weight: 283,300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801051
CAS-Nummer: 477857-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR