Tris[4-(trifluoromethyl)phenyl]phosphine 95+%

Lieferant: Apollo Scientific

Synonyme: Tris(4-trifluoromethylphenyl)phosphine, Tris(4-(trifluoromethyl)phenyl)phosphine

PC7919G-1G PC7919G-5G PC7919G-10G
APOSPC7919G-1GEA 36.4 EUR
APOSPC7919G-1G APOSPC7919G-5G APOSPC7919G-10G
Tris[4-(trifluoromethyl)phenyl]phosphine 95+%
Tris[4-(trifluoromethyl)phenyl]phosphine
Formel: C₂₁H₁₂F₉P
Molecular Weight: 466.28 g/mol
Schmelzpunkt: 73…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00058883
CAS-Nummer: 13406-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR