2-Chlor-3-fluorphenol
Lieferant: Apollo Scientific
|
Gefahr
|
PC7923-5G
APOSPC7923-5GEA
87
EUR
APOSPC7923-5G
2-Chlor-3-fluorphenol
2-Chlor-3-fluorphenol
Formel:
C₆H₄ClFO Molecular Weight: 146,5500 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08166413 CAS-Nummer: 863870-86-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...