1,1,3-Trifluoropropane 97%

Lieferant: Apollo Scientific

PC7926-1G
APOSPC7926-1GEA 176 EUR
APOSPC7926-1G
1,1,3-Trifluoropropane 97%
1,1,3-Trifluoropropane
Formel: C₃H₅F₃
Molecular Weight: 98,067 g/mol
Siedepunkt: 45 °C
Dichte: 1,182 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD07784186
CAS-Nummer: 24270-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR