2-Iod-6-nitro-4-(trifluormethyl)anilin 95+%
Lieferant: Apollo Scientific
Synonyme:
4-Amino-3-iodo-5-nitrobenzotrifluoride
PC7943-1G
PC7943-500MG
PC7943-5G
APOSPC7943-1GEA
270
EUR
APOSPC7943-1G
APOSPC7943-500MG
APOSPC7943-5G
2-Iod-6-nitro-4-(trifluormethyl)anilin 95+%
2-Iod-6-nitro-4-(trifluormethyl)anilin
Formel:
C₇H₄F₃IN₂O₂ Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD14560546 CAS-Nummer: 543740-74-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...