6-Chlor-2-fluorpurin

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-2-fluoro-9H-purine

PC7945-500MG
APOSPC7945-500MGEA 21.9 EUR
APOSPC7945-500MG
6-Chlor-2-fluorpurin
6-Chloro-2-fluoropurine
Formel: C₅H₂ClFN₄
Molecular Weight: 172,55 g/mol
Schmelzpunkt: 157…162 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02183557
CAS-Nummer: 1651-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR