3-Brom-2-chlor-5-(trifluormethyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

PC7968-1G
APOSPC7968-1GEA 58 EUR
APOSPC7968-1G
3-Brom-2-chlor-5-(trifluormethyl)phenylboronsäure 98%
3-Brom-2-chlor-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₄BBrClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800879
CAS-Nummer: 957120-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR