3-Bromo-5-(trifluoromethyl)pyridin-2-ol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-bromo-5-(trifluoromethyl)pyridin-2-ol, 3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 3-bromo-5-(trifluoromethyl)pyridine-2-ol
PC7971-1G
PC7971-5G
PC7971-10G
APOSPC7971-1GEA
21.9
EUR
APOSPC7971-1G
APOSPC7971-5G
APOSPC7971-10G
3-Bromo-5-(trifluoromethyl)pyridin-2-ol 98%
3-Bromo-5-(trifluoromethyl)pyridin-2-ol
Formel:
C₆H₃BrF₃NO Schmelzpunkt: 170…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02691223 CAS-Nummer: 76041-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...