6-Bromo-2-fluoro-3-(methoxycarbonyl)benzeneboronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
6-Bromo-2-fluoro-3-(methoxycarbonyl)phenylboronic acid
PC7972-1G
APOSPC7972-1GEA
256
EUR
APOSPC7972-1G
6-Bromo-2-fluoro-3-(methoxycarbonyl)benzeneboronic acid 98%
6-Bromo-2-fluoro-3-(methoxycarbonyl)benzeneboronic acid
Formel:
C₈H₇BBrFO₄ Molecular Weight: 276.852 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800873 CAS-Nummer: 957120-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...