tert-Butyl-2-chlor-4-fluorbenzoat

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-chloro-4-fluorobenzoate

PC7974-5G
APOSPC7974-5GEA 50.9 EUR
APOSPC7974-5G
tert-Butyl-2-chlor-4-fluorbenzoat
tert-Butyl-2-chlor-4-fluorbenzoat
Formel: C₁₁H₁₂ClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800898
CAS-Nummer: 911314-43-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR