Methyl 2-nitro-4-(trifluoromethyl)phenylacetate

Lieferant: Apollo Scientific
Achtung

PC7975-500MG
APOSPC7975-500MGEA 21.9 EUR
APOSPC7975-500MG
Methyl 2-nitro-4-(trifluoromethyl)phenylacetate
Methyl 2-nitro-4-(trifluoromethyl)phenylacetate
Formel: C₁₀H₈F₃NO₄
Molecular Weight: 263,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800854
CAS-Nummer: 13544-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR