2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid

Lieferant: Apollo Scientific

PC7979-1G
APOSPC7979-1GEA 176 EUR
APOSPC7979-1G
2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid
2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid
Formel: C₉H₉BrFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800913
CAS-Nummer: 951884-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR