2-Fluoro-3-methoxybenzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
PC7984-1G
APOSPC7984-1GEA
43.5
EUR
APOSPC7984-1G
2-Fluoro-3-methoxybenzyl alcohol
2-Fluoro-3-methoxybenzyl alcohol
Formel:
C₈H₉FO₂ Siedepunkt: 126…129 °C (5 mm Hg) Schmelzpunkt: 56…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09743596 CAS-Nummer: 178974-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...