Methyl-3-{[4-(trifluoromethyl)piperidino]methyl}benzenecarboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 3-{[4-(trifluoromethyl)piperidin-1-yl]methyl}benzoate

PC7994-1G
APOSPC7994-1GEA 176 EUR
APOSPC7994-1G
Methyl-3-{[4-(trifluoromethyl)piperidino]methyl}benzenecarboxylate
Methyl-3-{[4-(trifluoromethyl)piperidino]methyl}benzenecarboxylate
Formel: C₁₅H₁₈F₃NO₂
Molecular Weight: 301,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864681
CAS-Nummer: 1000339-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR