Methyl-4-{2-[4-(trifluoromethyl)piperidino]ethoxy}benzenecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-{2-[4-(trifluoromethyl)piperidin-1-yl]ethoxy}benzoate
PC7995-1G
APOSPC7995-1GEA
176
EUR
APOSPC7995-1G
Methyl-4-{2-[4-(trifluoromethyl)piperidino]ethoxy}benzenecarboxylate
Methyl-4-{2-[4-(trifluoromethyl)piperidino]ethoxy}benzenecarboxylate
Formel:
C₁₆H₂₀F₃NO₃ Molecular Weight: 331,34 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864682 CAS-Nummer: 1000339-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...