3-{2-[4-(Trifluoromethyl)piperidino]ethyl}benzenecarbaldehyde

Lieferant: Apollo Scientific

Synonyme: 3-{2-[4-(Trifluoromethyl)piperidin-1-yl]ethyl}benzaldehyde

PC7996-1G
APOSPC7996-1GEA 263 EUR
APOSPC7996-1G
3-{2-[4-(Trifluoromethyl)piperidino]ethyl}benzenecarbaldehyde
3-{2-[4-(Trifluoromethyl)piperidino]ethyl}benzenecarbaldehyde
Formel: C₁₅H₁₈F₃NO
Molecular Weight: 285,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864684
CAS-Nummer: 1000339-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR