4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde

Lieferant: Apollo Scientific

PC7998-1G
APOSPC7998-1GEA 176 EUR
APOSPC7998-1G
4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde
4-{2-[4-(Trifluoromethyl)piperidino]acetyl}-1H-pyrrole-2-carbaldehyde
Formel: C₁₃H₁₅F₃N₂O₂
Molecular Weight: 288,27 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09864685
CAS-Nummer: 1000339-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR