3-Chlor-4-fluor-5-nitrobenzotrifluorid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Chloro-2-fluoro-3-nitro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluoro-5-nitrobenzotrifluoride

PC8003-1G
APOSPC8003-1GEA 0 EUR
APOSPC8003-1G
3-Chlor-4-fluor-5-nitrobenzotrifluorid
3-Chlor-4-fluor-5-nitrobenzotrifluorid
Formel: C₇H₂ClF₄NO₂
Molecular Weight: 243,54 g/mol
Siedepunkt: 200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00674108
CAS-Nummer: 101646-02-0
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR