3-Chloro-2,5-difluoropyridine 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC8011-5G
APOSPC8011-5GEA
75.4
EUR
APOSPC8011-5G
3-Chloro-2,5-difluoropyridine 98%
3-Chloro-2,5-difluoropyridine
Formel:
C₅H₂ClF₂N Molecular Weight: 149,53 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998887 CAS-Nummer: 851179-00-5 UN: 2924 ADR: 3+8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...