2-Brom-N,N-dimethyl-5-fluorbenzamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-5-fluoro-N,N-dimethylbenzamide
PC8019-1G
PC8019-5G
PC8019-250MG
APOSPC8019-1GEA
190
EUR
APOSPC8019-1G
APOSPC8019-5G
APOSPC8019-250MG
2-Brom-N,N-dimethyl-5-fluorbenzamid 98%
2-Brom-N,N-dimethyl-5-fluorbenzamid
Formel:
C₉H₉BrFNO Molecular Weight: 246.0763 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800904 CAS-Nummer: 951884-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...